n-Hexadecyl-β-D-maltoside, Sigma-Aldrich®
Catalog # SIAL52318-100MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:566.72 g/mol
CAS Number:98064-96-1
Formula:C₂₈H₅₄O₁₁
MDL Number:MFCD01310880