Order Entry
Poland
ContactUsLinkComponent
2'-Deoxyguanosine-5'-triphosphate trisodium salt (dGTP-Na3) 100 mmol/l in aqueous solution, Sigma-Aldrich®
2'-Deoxyguanosine-5'-triphosphate trisodium salt (dGTP-Na3) 100 mmol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIALD5038-.1MMO
Supplier:  Merck
CAS Number:  
2'-Deoxyguanosine-5'-triphosphate trisodium salt (dGTP-Na3) 100 mmol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIALD5038-.1MMO
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    0,1 mmol
  • Seq:
    9155

Specifications

About this item

 :
 :573.12 g/mol
EINECS:300-026-5
CAS Number:93919-41-6
Formula:C₁₀H₁₃N₅Na₃O₁₃P₃
MDL Number:MFCD00080337