Order Entry
Poland
ContactUsLinkComponent
Pig L-Lactate dehydrogenase (from Heart)
Pig L-Lactate dehydrogenase (from Heart)
Catalog # SIALL7525-12.5KU
Supplier:  Merck
CAS Number:  
Pig L-Lactate dehydrogenase (from Heart)
Catalog # SIALL7525-12.5KU
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    12,5 KU
  • CAS No.:
    9001-60-9
  • Source:
    Heart
  • Species:
    Pig
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00131466
  • Enzyme name:
    L-Lactate dehydrogenase
  • Enzyme synonyms:
    LDH

Specifications

About this item