Order Entry
Poland
ContactUsLinkComponent
Leupeptin hemisulphate
Leupeptin hemisulphate
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Leupeptin hemisulphate
Supplier:  ENZO LIFE SCIENCES

Specifications

  • CAS No.:
    103476-89-7
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00037012
  • Protein synonyms:
    (Ac-Leu-Leu-Arg-al hemisulphate) monohydrate
  • Protein/peptide name:
    Leupeptin hemisulphate
  • Molecular formula:
    C₄₀H₇₈N₁₂O₁₂S

Specifications

Frequently Bought Together