Order Entry
Poland
Orders LinkContactUsLinkComponent
E-64

E-64

Supplier:   Cayman Chemical
E-64
Supplier:  Cayman Chemical
Certificates

4 Options Available Below

Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CAYM10007963-5
  • CAYM10007963-10
  • CAYM10007963-25
  • CAYM10007963-50
  • Protein/peptide name:
  • E-64
  • E-64
  • E-64
  • E-64
  • Protein synonyms:
  • 3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
  • 3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
  • 3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
  • 3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
  • CAS No.:
  • 66701-25-5
  • 66701-25-5
  • 66701-25-5
  • 66701-25-5
  • MDL No.:
  • MFCD00080261
  • MFCD00080261
  • MFCD00080261
  • MFCD00080261
  • Molecular formula:
  • C₁₅H₂₇N₅O₅
  • C₁₅H₂₇N₅O₅
  • C₁₅H₂₇N₅O₅
  • C₁₅H₂₇N₅O₅
  • Storage conditions:
  • Freezer
  • Freezer
  • Freezer
  • Freezer

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 393 zł
    393 zł
    1 * 5 mg
       393 zł

    Specifications:
    More
    Specifications
    Cat. No.
    CAYM10007963-5
    Protein/peptide name
    E-64
    Protein synonyms
    3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
    CAS No.
    66701-25-5
    MDL No.
    MFCD00080261
    Molecular formula
    C₁₅H₂₇N₅O₅
    Storage conditions
    Freezer
  • Glass bottle for solids
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 704 zł
    704 zł
    1 * 10 mg
       704 zł

    Specifications:
    More
    Specifications
    Cat. No.
    CAYM10007963-10
    Protein/peptide name
    E-64
    Protein synonyms
    3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
    CAS No.
    66701-25-5
    MDL No.
    MFCD00080261
    Molecular formula
    C₁₅H₂₇N₅O₅
    Storage conditions
    Freezer
  • Glass bottle for solids
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 580 zł
    1 580 zł
    1 * 25 mg
       1 580 zł

    Specifications:
    More
    Specifications
    Cat. No.
    CAYM10007963-25
    Protein/peptide name
    E-64
    Protein synonyms
    3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
    CAS No.
    66701-25-5
    MDL No.
    MFCD00080261
    Molecular formula
    C₁₅H₂₇N₅O₅
    Storage conditions
    Freezer
  • Glass bottle for solids
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 2 940 zł
    2 940 zł
    1 * 50 mg
       2 940 zł

    Specifications:
    More
    Specifications
    Cat. No.
    CAYM10007963-50
    Protein/peptide name
    E-64
    Protein synonyms
    3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
    CAS No.
    66701-25-5
    MDL No.
    MFCD00080261
    Molecular formula
    C₁₅H₂₇N₅O₅
    Storage conditions
    Freezer
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.