Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Malt extract, Millipore®

Glass bottle for solids, 500 g
Catalog # SIAL70167-500G
Supplier:   Merck
Malt extract, Millipore®
Catalog # SIAL70167-500G
Supplier:  Merck
CAS Number:  8002-48-0
Malt extract, Millipore®

Glass bottle for solids, 500 g

CAS Number:  8002-48-0
Price
683 zł
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:8002-48-0
  • MDL Number:MFCD00131570
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:232-310-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications