3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl) (synthetic), proszek
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7411-49-6
:
Formula:C₁₂H₁₈Cl₄N₄
MDL Number:MFCD00012969
:360.11 g/mol
EINECS:231-018-9
Frequently Bought Together