2-Mercaptoethanesulphonic acid sodium salt (mesna) for synthesis, Sigma-Aldrich®
: Merck
:19767-45-4
:
:C₂H₅NaO₃S₂
:MFCD00007535
:M-Clarity™ quality level = MQ200
:164.18 g/mol
:243-285-9
- Solubility:(20 °C) soluble
- Molar Mass:164.18 g/mol
Frequently Bought Together