Order Entry
Poland
ContactUsLinkComponent
1-Naphthyl phosphate monosodium salt monohydrate
Irritant
1-Naphthyl phosphate monosodium salt monohydrate
Catalog # MOLEM69305565
Supplier:  Molekula
CAS Number:  
Irritant
1-Naphthyl phosphate monosodium salt monohydrate
Catalog # MOLEM69305565
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :264.15 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:81012-89-7
Formula:C₁₀H₈NaO₄P·1H₂O
MDL Number:MFCD00150615