Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Azauracil
6-Azauracil, Sigma-Aldrich®
6-Azauracil, Sigma-Aldrich®
6-Azauracil, Sigma-Aldrich®
SIALA1757-5G
:
Merck
:
6-Azauracil, Sigma-Aldrich®
SIALA1757-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
113.08 g/mol
:
207-318-0
:
461-89-2
:
C₃H₃N₃O₂
:
MFCD00006456