Order Entry
Poland
ContactUsLinkComponent
Avidin (from egg white), MP Biomedicals
Avidin (from egg white), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Avidin (from egg white), MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    1405-69-2
  • Source:
    Egg white
  • Storage conditions:
    −20 °C
  • Form:
    Crystalline Powder
  • MDL No.:
    MFCD00130572
  • Protein/peptide name:
    Avidin
  • Colour:
    White

Specifications

Frequently Bought Together