Order Entry
Poland
ContactUsLinkComponent
 
Acetonyltriphenylphosphonium chloride for synthesis, Sigma-Aldrich®
  8.41028.0025
 :  Merck
 :  
Acetonyltriphenylphosphonium chloride for synthesis, Sigma-Aldrich®
  8.41028.0025
 :  Merck
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Melting Point:
    243 - 245 °C
  • Molar Mass:
    354.81 g/mol
  • Synonyms:
    2-Oxopropyl triphenylphosphonium chloride
  • Hill Formula:
    C21H20ClOP

 

 

 :
 :354.82 g/mol
 :243…245 °C
 :M-Clarity™ quality level = MQ200
 :Ambient
 :214-974-1
 :1235-21-8
 :C₂₁H₂₀ClOP
 :MFCD00011813