Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Bacitracin

Glass bottle for solids, 25 g
Catalog # MOLE64862649-25G
Marketsource
Supplier:   Molekula
Bacitracin
Catalog # MOLE64862649-25G
Supplier:  Molekula
Marketsource
CAS Number:  1405-87-4
Bacitracin

Glass bottle for solids, 25 g

CAS Number:  1405-87-4
Price
1 425 zł
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:1405-87-4
  • Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
  • MDL Number:MFCD00062640
  • Molecular Weight:1421.60 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.