Bovine aprotinin (from Lung)
Catalog # USBIA2300-05-50
Supplier: US Biological
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:50 mg
- CAS No.:9087-70-1
- Species:Bovine
- Storage conditions:Refrigerator
- MDL No.:MFCD00130541
- Protein synonyms:Trypsin inhibitor, pancreatic basic
- Protein/peptide name:Aprotinin
- Molecular formula:C₂₈₄H₄₃₂N₈₄O₇₉S₇