Order Entry
Poland
ContactUsLinkComponent
Pluronic® F-68 10% in aqueous solution, BioReagent cell culture reagent, sterile filtered, Sigma-Aldrich®
Pluronic® F-68 10% in aqueous solution, BioReagent cell culture reagent, sterile filtered, Sigma-Aldrich®
Catalog # SIALP5556-100ML
Supplier:  Merck
CAS Number:  
Pluronic® F-68 10% in aqueous solution, BioReagent cell culture reagent, sterile filtered, Sigma-Aldrich®
Catalog # SIALP5556-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance (Turbidity):
    Clear
  • Appearance (Form):
    Solution
  • pH:
    6.1 - 7.1
  • Osmolality:
    ≤200 mOs/kg
  • Sterility by current EP/USP:
    Passes
  • Endotoxin Level:
    ≤10 EU/ml
  • Concentration by CDET:
    9.0 - 11.0 %
  • Cell Culture Testing - MTT:
    Pass

Specification Test Results

About this item

 :
 :8400
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
CAS Number:9003-11-6
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
MDL Number:MFCD00082049