Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:71-44-3
:
Formula:C₁₀H₂₆N₄
MDL Number:MFCD00008215
:202.35 g/mol
:M-Clarity™ quality level = MQ200
EINECS:200-754-2
Frequently Bought Together