
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) trihydrate
Supplier: Molekula
Certificates
About this item
CAS Number:606-68-8
:
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂·3H₂O
MDL Number:MFCD00036200
EINECS:210-123-3
:GHS07
Storage Temperature:Freezer
Frequently Bought Together