Order Entry
Poland
ContactUsLinkComponent
Cresol red
Cresol red
Catalog # MOLEM79737710
Supplier:  Molekula
CAS Number:  
Cresol red
Catalog # MOLEM79737710
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :382.43 g/mol
Storage Temperature:Ambient
EINECS:217-064-2
CAS Number:1733-12-6
Formula:C₂₁H₁₈O₅S
MDL Number:MFCD00005878