Cresol red
Supplier: Merck
Certificates
About this item
CAS Number:1733-12-6
:
Formula:C₂₁H₁₈O₅S
MDL Number:MFCD00005878
:382.43 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:217-064-2
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:290 °C decomposes
- Molar Mass:382.43 g/mol
- Bulk Density:600 - 620 kg/m3
Specification Test Results
Frequently Bought Together