Nonidet® P 40 Substitute (NP-40) 10% in aqueous solution
:9016-45-9
:
:MFCD00132851
:500-024-6
Frequently Bought Together
Water, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals
Ethylene glycol ≥99% (by GC), pure
: PanReac AppliChem
Glycerol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 1.000 mL
: PanReac AppliChem
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
: Honeywell Chemicals
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
: Thermo Fisher Scientific