ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form), Sigma-Aldrich®
Catalog # SIALN8160-15VL
Supplier: Merck
CAS Number:
Specifications
- Pk:15 Vial
- Seq:0277
Specifications
About this item
:
:765.39 g/mol
Melting Pt:175 to 178 °C
EINECS:214-664-6
CAS Number:1184-16-3
Formula:C₂₁H₂₇N₇NaO₁₇P₃
MDL Number:MFCD00036973