β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®
Catalog # SIALN9535-15VL
Supplier: Merck
CAS Number:
Specifications
- Pk:15 Vial
- Seq:0277
Specifications
About this item
:
:709.41 g/mol
EINECS:210-123-3
CAS Number:606-68-8
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number:MFCD00036200