Order Entry
Poland
ContactUsLinkComponent
Polyvinylpyrrolidone, M.W. ∼40000 ≥99%, high purity (K30)
Polyvinylpyrrolidone, M.W. ∼40000 ≥99%, high purity (K30)
Catalog # 0507-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Polyvinylpyrrolidone, M.W. ∼40000 ≥99%, high purity (K30)
Catalog # 0507-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Heavy Metals:
    <0.001%
  • pH (5%, Water) @25 °C:
    3 – 7
  • Purity (Dry Basis):
    >99%

Specification Test Results

About this item

 :
Storage Temperature:Ambient
CAS Number:9003-39-8
Formula:(C₆H₉NO)n
MDL Number:MFCD01076626