Order Entry
Poland
ContactUsLinkComponent
3,3-Dimethylglutaric anhydride, Sigma-Aldrich®
3,3-Dimethylglutaric anhydride, Sigma-Aldrich®
Catalog # SIALD159808-100G
Supplier:  Merck
CAS Number:  
3,3-Dimethylglutaric anhydride, Sigma-Aldrich®
Catalog # SIALD159808-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :142.15 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:223-998-1
CAS Number:4160-82-1
Formula:C₇H₁₀O₃
MDL Number:MFCD00006684