Order Entry
Poland
ContactUsLinkComponent
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-18, SB-18)
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-18, SB-18)
Catalog # MOLE42559860-25G
Supplier:  Molekula
CAS Number:  
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-18, SB-18)
Catalog # MOLE42559860-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Storage Temperature:Ambient
EINECS:236-124-9
CAS Number:13177-41-8
Formula:C₂₃H₄₉NO₃S
MDL Number:MFCD00043218