Siarczan(VI) sodu 0.25 mol/l in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
Synonyms: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt
CAS Number:7757-82-6
:
Formula:Na₂SO₄
MDL Number:MFCD00003504
:142.04 g/mol
Storage Temperature:Temperatura otoczenia
EINECS:231-820-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together