Sodium chloride 200 mg/l in aqueous solution volumetric solution
Catalog # CHMP817935120.1000
Supplier: CHEMPUR
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
Synonyms: Saline, NaCl, Sea salt, Halite
:
:58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477