Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium chloride
Sodium chloride 50 mg/l in aqueous solution volumetric solution
Sodium chloride 50 mg/l in aqueous solution volumetric solution
Sodium chloride 50 mg/l in aqueous solution volumetric solution
CHMP817935121.1000
:
CHEMPUR
:
Sodium chloride 50 mg/l in aqueous solution volumetric solution
CHMP817935121.1000
:
CHEMPUR
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
58.44 g/mol
:
Ambient
:
231-598-3
:
7647-14-5
:
NaCl
:
MFCD00003477