
Lithium chloride 1 mol/l in ethanol volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7447-41-8
:
Formula:LiCl
MDL Number:MFCD00011078
:42.39 g/mol
Storage Temperature:Ambient
EINECS:231-212-3
UN:1170
ADR:3,II
:GHS02
Frequently Bought Together