Lithium chloride 0.1 mol/l in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7447-41-8
:
Formula:LiCl
MDL Number:MFCD00011078
:42.39 g/mol
Storage Temperature:Ambient
EINECS:231-212-3
Frequently Bought Together