Disodium oxalate 0.00125 mol/l (0.0025 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:62-76-0
:
Formula:Na₂C₂O₄
MDL Number:MFCD00012465
:134 g/mol
Storage Temperature:Ambient
EINECS:200-550-3
Frequently Bought Together