Sodium carbonate 0.5 mol/l (1 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:497-19-8
:
Formula:Na₂CO₃
MDL Number:MFCD00003494
:105.99 g/mol
Storage Temperature:Ambient
EINECS:207-838-8
Frequently Bought Together