0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Head Protection
Safety Helmets, customised
Safety helmet with print DIW, ECO 1100
Safety helmet with print DIW, ECO 1100
Safety helmet with print DIW, ECO 1100
Catalog # CENUS01EBCP-DIW1
Supplier:
CENTURION SAFETY PRODUCTS
Expand image
Safety helmet with print DIW, ECO 1100
Catalog # CENUS01EBCP-DIW1
Supplier:
CENTURION SAFETY PRODUCTS
Specifications
Colour:
Blue
Description:
Safety helmet, DIW ECO 1100, with print
Pk:
1
Specifications
About this item
Product Details
Product Details