
Tetracycline hydrochloride, Sigma-Aldrich®
Catalog # SIALT7660-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:480.9 g/mol
:M-Clarity™ quality level = MQ200
EINECS:200-593-8
:GHS07
Storage Temperature:Ambient
CAS Number:64-75-5
Formula:C₂₂H₂₅ClN₂O₈
MDL Number:MFCD00078142