

Chlortetracycline hydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:64-72-2
:
Formula:C₂₂H₂₃ClN₂O₈·HCl
MDL Number:MFCD26142699
:515.34 g/mol
EINECS:200-591-7
:GHS06
:GHS08
Storage Temperature:Refrigerator
Frequently Bought Together