Order Entry
Poland
ContactUsLinkComponent
Chlortetracycline hydrochloride
Toxic
Health
Chlortetracycline hydrochloride
Catalog # PROOICRS0249
CAS Number:  
Toxic
Health
Chlortetracycline hydrochloride
Catalog # PROOICRS0249
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 mg
  • Seq:
    2530

Specifications

About this item

 :
 :515.34 g/mol
EINECS:200-591-7
 :GHS06
 :GHS08
Storage Temperature:Refrigerator
CAS Number:64-72-2
Formula:C₂₂H₂₃ClN₂O₈·HCl
MDL Number:MFCD26142699