Order Entry
Poland
ContactUsLinkComponent
Chicken lysozyme (Muramidase) (from egg white)
Chicken lysozyme (Muramidase) (from egg white)
Catalog # 1.05281.0010
CAS Number:  
Chicken lysozyme (Muramidase) (from egg white)
Catalog # 1.05281.0010
CAS Number:  

Specifications

  • Pk:
    10 g
  • CAS No.:
    9001-63-2
  • Source:
    Egg white
  • Species:
    Chicken
  • Seq:
    02
  • Storage conditions:
    Freezer
  • Form:
    White
  • Enzyme name:
    Lysozyme (Muramidase)
  • Enzyme activity:
    ≥30 000 FIP-U/mg
  • pH:
    3,5

Specifications

About this item