Order Entry
Poland
ContactUsLinkComponent
Transport trolleys for baskets, EURO-STANDARD | Trolleys
Transport trolleys for baskets, EURO-STANDARD
Catalog # BOCH17333
Supplier:  BOCHEM
undefined
Transport trolleys for baskets, EURO-STANDARD
Catalog # BOCH17333
Supplier:  BOCHEM
Specifications
  • Description:
    Transport cart
  • W×D×H:
    610×465×1360 mm
  • Pk:
    1
Specifications
About this item
Transport cart made of stainless steel used for transport baskets.
  • Five hanging ties
  • Two pushing handles
  • Two guide and fixed rolls, two with lock
  • Enclosure of rolls, zincked
  • Excess length of enclosure