Order Entry
Poland
ContactUsLinkComponent
Hygromycin B 50 mg/ml in solution, Ultra Pure Grade
Hygromycin B 50 mg/ml in solution, Ultra Pure Grade
Catalog # K547-20ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hygromycin B 50 mg/ml in solution, Ultra Pure Grade
Catalog # K547-20ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    20 mL
  • Seq:
    2305

Specifications

About this item

 :
 :527.5 g/mol
CAS Number:31282-04-9
Formula:C₂₀H₃₇N₃O₁₃
MDL Number:MFCD06795479