Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Ectoine, Sigma-Aldrich®

Supplier:   Merck
Ectoine, Sigma-Aldrich®
Supplier:  Merck
Certificates
Ectoine, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  (S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
  • CAS Number:96702-03-3
  • Formula:C₆H₁₀N₂O₂
  • MDL Number:MFCD03419286
  • Molecular Weight:142.1558 g/mol
  •  :M-Clarity™ quality level = MQ200
  • EINECS:431-910-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 270 zł
    1 270 zł
    1 * 25 mg
       1 270 zł

  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 4 250 zł
    4 250 zł
    1 * 100 mg
       4 250 zł