Order Entry
Poland
Orders LinkContactUsLinkComponent
α-Cyclodextrin, Sigma-Aldrich®

α-Cyclodextrin, Sigma-Aldrich®

Supplier:   Merck
α-Cyclodextrin, Sigma-Aldrich®
Supplier:  Merck
Certificates
α-Cyclodextrin, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  •  :M-Clarity™ quality level = MQ300
  • EINECS:233-007-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 204 zł
    204 zł
    1 * 1 g
       204 zł

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 330 zł
    330 zł
    1 * 5 g
       330 zł

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 400 zł
    1 400 zł
    1 * 25 g
       1 400 zł