Order Entry
Poland
Orders LinkContactUsLinkComponent
α-Cyclodextrin, Sigma-Aldrich®

α-Cyclodextrin, Sigma-Aldrich®

Supplier:   Merck
α-Cyclodextrin, Sigma-Aldrich®
Supplier:  Merck
Certificates
α-Cyclodextrin, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  •  :M-Clarity™ quality level = MQ200
  • EINECS:233-007-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 365 zł
    365 zł
    1 * 5 g
       365 zł

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 200 zł
    1 200 zł
    1 * 25 g
       1 200 zł

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 3 560 zł
    3 560 zł
    1 * 100 g
       3 560 zł

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.