Potassium nitrate 2 mol/l in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7757-79-1
:
Formula:KNO₃
MDL Number:MFCD00011409
:101.1 g/mol
Storage Temperature:Ambient
EINECS:231-818-8
Frequently Bought Together