2,6-Dichloroquinone-4-chloroimide, GR Reag. Ph. Eur., Supelco®
Catalog # 1.03037.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay (iodometric):≥ 98
- Identity (IR-spectrum):passes test
- Spec. Absorptivity A 1%/1cm (312 nm; 0,001 %; 1 cm; ethanol):≥ 800
- Melting point:65 - 68
- Hazard Class:4.1
- Hazard Class Description:Flammable solid
Specification Test Results
About this item
:
ADR:4.1,
:M-Clarity™ quality level = MQ100
EINECS:202-937-2
UN:3224
:GHS02
:GHS07
Storage Temperature:Refrigerator
CAS Number:101-38-2
Formula:C₆H₂Cl₃NO
MDL Number:MFCD00001611