Nickel(II) carbonate basic hydrate, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Nickel(II) carbonate hydroxide hydrate, Basic nickel carbonate
- CAS Number:12607-70-4
- Formula:~NiCO₃ · 2Ni(OH)₂ · 4H₂O
- MDL Number:MFCD00016262
- Molecular Weight:304.11 g/mol (anhy)
- EINECS:235-715-9
2 Options Available Below
Product Details & Documents
Synonyms: Nickel(II) carbonate hydroxide hydrate, Basic nickel carbonate
- CAS Number:12607-70-4
- Formula:~NiCO₃ · 2Ni(OH)₂ · 4H₂O
- MDL Number:MFCD00016262
- Molecular Weight:304.11 g/mol (anhy)
- EINECS:235-715-9
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



