4-(2-Chloroethyl)morpholine hydrochloride
Catalog # MOLEM60023687
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:186.08 g/mol
EINECS:222-881-2
UN:2923
ADR:8,III
CAS Number:3647-69-6
Formula:C₆H₁₃Cl₂NO
MDL Number:MFCD00012797