Order Entry
Poland
ContactUsLinkComponent
Ethylmagnesium bromide 1 mol/l in tert-butyl methyl ether, Sigma-Aldrich®
Ethylmagnesium bromide 1 mol/l in tert-butyl methyl ether, Sigma-Aldrich®
Catalog # SIAL345105-100ML
Supplier:  Merck
CAS Number:  
Ethylmagnesium bromide 1 mol/l in tert-butyl methyl ether, Sigma-Aldrich®
Catalog # SIAL345105-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :133.27 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:213-127-3
CAS Number:925-90-6
Formula:C₂H₅BrMg
MDL Number:MFCD00000043