Order Entry
Poland
ContactUsLinkComponent
Holmium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Oxidizing
Irritant
Holmium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Catalog # 40575.NC
CAS Number:  
Oxidizing
Irritant
Holmium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Catalog # 40575.NC
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

About this item

CAS Number:14017-54-0
 :
Formula:Cl₃HoO₁₂
 :463.28
Density:1.57
Storage Temperature:Ambient
EINECS:237-839-9
UN:3211
ADR:5.1,II
 :GHS03
 :GHS07
MDL Number:MFCD00064643