Holmium(III) perchlorate 40% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:14017-54-0
:
Formula:Cl₃HoO₁₂
MDL Number:MFCD00064643
:463.28
:M-Clarity™ quality level = MQ100
EINECS:237-839-9
Frequently Bought Together