
Sulphuric acid 0.2 mol concentrated aqueous solution volumetric solution
Catalog # CHMP165750642.0001
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589